missing translation for 'onlineSavingsMsg'
Learn More

HCAP-G Antibody, Novus Biologicals™

Product Code. 18363076 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363076 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363076 Supplier Novus Biologicals Supplier No. NBP312992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HCAP-G Polyclonal antibody specifically detects HCAP-G in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCAP-G
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias CAP-G, CAPGHCAP-G, CHCG, chromosome condensation protein G, Chromosome-associated protein G, condensin complex subunit 3, Condensin subunit CAP-G, FLJ12450, hCAP-G, Melanoma antigen NY-MEL-3, MGC126525, Non-SMC condensin I complex subunit G, non-SMC condensin I complex, subunit G, NYMEL3, NY-MEL-3, XCAP-G homolog
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HCAP-G. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cellular Markers, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 64151
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.