missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HC-067047, MedChemExpress
Description
HC-067047 is a potent and selective TRPV4 antagonist and reversibly inhibits currents through the human, rat, and mouse TRPV4 orthologs with IC50 values of 48 nM, 133 nM, and 17 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | HC-067047 |
| CAS | 883031-03-6 |
| Color | Light Nude |
| Purity Grade Notes | Research |
| Molecular Formula | C26H28F3N3O2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 50 mg/mL (106.04 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(C)N(CCCN2CCOCC2)C(C3=CC=CC=C3)=C1)NC4=CC=CC(C(F)(F)F)=C4 |
| Molecular Weight (g/mol) | 471.51 |
| Formula Weight | 471.51 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?