missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HC 067047

Product Code. 18045232
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18045232 50 mg 50mg
18015432 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18045232 Supplier Tocris Bioscience™ Supplier No. 4100/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective TRPV4 antagonist

HC 067047 is a potent and selective TRPV4 antagonist. Reversibly inhibits currents through mouse, human and rat TRPV4 orthologs (IC50 values are 17, 48 and 133 nM respectively). Also inhibits the endogenous TRPV4-mediated response to 4α-PDH (IC50 = 22 nM). Selective for TRPV4 over TRPV1, TRPV2, TRPV3 and TRPM8 channels.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methyl-1-[3-(4-morpholinyl)propyl]-5-phenyl-N-[3-(trifluoromethyl)phenyl]-1H-pyrrole-3-carboxamide
CAS 883031-03-6
Quantity 50 mg
Target TRPV Antagonists
Molecular Formula C26H28F3N3O2
Purity 0.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.