missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBZ Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67878-1-IG-20UL
This item is not returnable.
View return policy
Description
The HBZ antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HBZ. This antibody recognizes human antigen. The HBZ antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| HBZ | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3050 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
| Western Blot, ELISA | |
| 1F9C2 | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 14-16 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction