missing translation for 'onlineSavingsMsg'
Learn More

HBX 19818, MedChemExpress

Product Code. 16467356
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467356 100 mg 100mg
16459857 5 mg 5mg
16447388 10 mg 10mg
16443417 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467356 Supplier Medchem Express Supplier No. HY17540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HBX 19818 is a specific inhibitor of ubiquitin-specific protease 7 (USP7), with an IC50 of 28.1 μM.

HBX 19818 is a specific inhibitor of ubiquitin-specific protease 7 (USP7), with an IC50 of 28.1 μM.

Tekniske data

Chemical Name or Material HBX 19818
Physical Form Solid
CAS 1426944-49-1
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C25H28ClN3O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 20 mg/mL (47.40 mM; Need ultrasonic)
SMILES CN(CCCNC(C1=CC=C(N=C(CCCC2)C2=C3Cl)C3=C1)=O)CC4=CC=CC=C4
Molecular Weight (g/mol) 421.96
Formula Weight 421.96
Percent Purity 98.39%
Grade Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.