missing translation for 'onlineSavingsMsg'
Learn More

HBTU, TRC

Product Code. 30402878
Change view
Click to view available options
Quantity:
1 g
5 g
Product Code. Quantity
30402878 5 g
30390346 1 g
2 options
This item is not returnable. View return policy

Product Code. 30402878

Brand: Toronto Research Chemicals TRCH1075005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94790-37-1
Molecular Formula C11H16F6N5OP
Molecular Weight (g/mol) 379.24
Synonym 1-[Bis(dimethylamino)methylene]-1H-benzotriazolium Hexafluorophosphate(1-) 3-Oxide, N-HBTU
IUPAC Name [benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)On1nnc2ccccc12.F[P-](F)(F)(F)(F)F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C11H16N5O.F6P/c1-14(2)11(15(3)4)17-16-10-8-6-5-7-9(10)12-13-16;1-7(2,3,4,5)6/h5-8H,1-4H3;/q+1;-1
Formula Weight 379.0997
Chemical Name or Material HBTU
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.