missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBsAg Antibody (Hs33), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-73138AF647
This item is not returnable.
View return policy
Description
HBsAg Monoclonal antibody specifically detects HBsAg in Virus samples. It is validated for Sandwich ELISA
Specifications
| HBsAg | |
| Monoclonal | |
| Alexa Fluor 647 | |
| HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant HBsAg of ayw subtype. | |
| 0.2 mL | |
| Virology Bacteria and Parasites | |
| 944569 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Sandwich ELISA | |
| Hs33 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction