missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBsAg Antibody (HB3), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
£453.00
Specifications
| Antigen | HBsAg |
|---|---|
| Clone | HB3 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 450 SE |
Description
HBsAg Monoclonal antibody specifically detects HBsAg in Virus samples. It is validated for ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| HBsAg | |
| ELISA | |
| mFluor Violet 450 SE | |
| Mouse | |
| Cell Biology, Virology Bacteria and Parasites | |
| 50mM Sodium Borate | |
| 944569 | |
| IgG1 | |
| Ion exchange chromatography |
| HB3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
| Purified HBsAg from human plasma. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title