missing translation for 'onlineSavingsMsg'
Learn More

HBsAg Antibody (HB3), DyLight 594, Novus Biologicals™

Product Code. 18403133 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403133 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403133 Supplier Novus Biologicals Supplier No. NB500474DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HBsAg Monoclonal antibody specifically detects HBsAg in Virus samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBsAg
Applications ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone HB3
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
Host Species Mouse
Immunogen Purified HBsAg from human plasma.
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 944569
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.