missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBS1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10359-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody specifically recognizes the 75 kDa human HBS1L protein.
This gene encodes a member of the GTP-binding elongation factor family. It is expressed in multiple tissues with the highest expression in heart and skeletal muscle. The intergenic region of this gene and the MYB gene has been identified to be a quantitative trait locus (QTL) controlling fetal hemoglobin level, and this region influnces erythrocyte, platelet, and monocyte counts as well as erythrocyte volume and hemoglobin content. DNA polymorphisms at this region associate with fetal hemoglobin levels and pain crises in sickle cell disease. A single nucleotide polymorphism in exon 1 of this gene is significantly associated with severity in beta-thalassemia/Hemoglobin E. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.Specifications
| HBS1L | |
| Polyclonal | |
| Unconjugated | |
| HBS1L | |
| EF 1a, ERFS, HBS1, HBS1 like (S. cerevisiae), HBS1 like protein, HBS1L, HSPC276, KIAA1038 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10767, 293408, 56422 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q69ZS7, Q6AXM7, Q9Y450 | |
| Hbs1l | |
| HBS1L Fusion Protein Ag0388 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction