missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HBS1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556240
This item is not returnable.
View return policy
Description
Immunogen sequence: CITGKIEAGY IQTGDRLLAM PPNETCTVKG ITLHDEPVDW AAAGDHVSLT LVGMDIIKIN VGCIFCGPKV PIKACTRF Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
This gene encodes a member of the GTP-binding elongation factor family. It is expressed in multiple tissues with the highest expression in heart and skeletal muscle. The intergenic region of this gene and the MYB gene has been identified to be a quantitative trait locus (QTL) controlling fetal hemoglobin level, and this region influnces erythrocyte, platelet, and monocyte counts as well as erythrocyte volume and hemoglobin content. DNA polymorphisms at this region associate with fetal hemoglobin levels and pain crises in sickle cell disease. A single nucleotide polymorphism in exon 1 of this gene is significantly associated with severity in beta-thalassemia/Hemoglobin E. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.
Specifications
| HBS1L | |
| Polyclonal | |
| Unconjugated | |
| HBS1L | |
| 2810035F15Rik; AI326327; EF-1a; eRF3 family member; eRF3c; ERF3-similar protein; ERFS; eukaryotic release factor 3; Hbs1; HBS1 like translational GTPase; HBS1L; Hbs1-like (S. cerevisiae); HBS1-like protein; HBS1-like translational GTPase; Hsp70 subfamily B suppressor 1-like protein; HSPC276; Kiaa1038 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10767 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y450 | |
| HBS1L | |
| Recombinant protein corresponding to Human HBS1L. Recombinant protein control fragment (Product #RP-94689). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction