missing translation for 'onlineSavingsMsg'
Learn More

HBS1L Mouse, anti-Human, CoraLite Plus 488, Clone: 1A10C4, Proteintech

Mouse Monoclonal Antibody

£135.00 - £375.00

Specifications

Antigen HBS1L
Clone 1A10C4
Concentration 1000 μg/mL
Applications Immunofluorescence
Conjugate CoraLite Plus 488
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18009655
View Documents
Proteintech
CL488-68146-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18080316
View Documents
Proteintech
CL488-68146-100UL
100 μL
£375.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The HBS1L antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HBS1L. This antibody recognizes human antigen. The HBS1L antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.

Specifications

Specifications

HBS1L
1000 μg/mL
CoraLite Plus 488
Mouse
10767
IgG1
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
1A10C4
Immunofluorescence
Liquid
Human
DKFZp686L13262, EF 1a, ERFS, HBS1, HBS1 like (S. cerevisiae), HBS1 like protein, HBS1L, HSPC276, KIAA1038
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.