missing translation for 'onlineSavingsMsg'
Learn More

HBS1L Antibody, Novus Biologicals™

Product Code. 18372119 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372119 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372119 Supplier Novus Biologicals Supplier No. H00010767B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

HBS1L Polyclonal antibody specifically detects HBS1L in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBS1L
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_006611.1
Gene Alias DKFZp434g247, ERF3-similar protein, ERFSDKFZp686L13262, HBS1 (S. cerevisiae)-like, HBS1KIAA1038EF-1a, HBS1-like (S. cerevisiae), HBS1-like protein, Hsp70 subfamily B suppressor 1-like protein, HSPC276
Host Species Mouse
Immunogen HBS1L (NP_006611.1, 1 a.a. - 684 a.a.) full-length human protein. MARHRNVRGYNYDEDFEDDDLYGQSVEDDYCISPSTAAQFIYSRRDKPSVEPVEEYDYEDLKESSNSVSNHQLSGFDQARLYSCLDHMREVLGDAVPDEILIEAVLKNKFDVQKALSGVLEQDRVQSLKDKNEATVSTGKIAKGKPVDSQTSRSESEIVPKVAKMTVSGKKQTMGFEVPGVSSEENGHSFHTPQKGPPIEDAIASSDVLETASKSANPPHTIQASEEQSSTPAPVKKSGKLRQQIDVKAELEKRQGGKQLLNLVVIGHVDAGKSTLMGHMLYLLGNINKRTMHKYEQESKKAGKASFAYAWVLDETGEERERGVTMDVGMTKFETTTKVITLMDAPGHKDFIPNMITGAAQADVAVLVVDASRGEFEAGFETGGQTREHGLLVRSLGVTQLAVAVNKMDQVNWQQERFQEITGKLGHFLKQAGFKESDVGFIPTSGLSGENLITRSQSSELTKWYKGLCLLEQIDSFKPPQRSIDKPFRLCVSDVFKDQGSGFCITGKIEAGYIQTGDRLLAMPPNETCTVKGITLHDEPVDWAAAGDHVSLTLVGMDIIKINVGCIFCGPKVPIKACTRFRARILIFNIEIPITKGFPVLLHYQTVSEPAVIKRLISVLNKSTGEVTKKKPKFLTKGQNALVELQTQRPIALELYKDFKELGRFMLRYGGSTIAAGVVTEIKE
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10767
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.