missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBP1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06527JF549
This item is not returnable.
View return policy
Description
HBP1 Polyclonal antibody specifically detects HBP1 in Human samples. It is validated for ELISA
Specifications
| HBP1 | |
| Polyclonal | |
| ELISA | |
| FLJ16340, High mobility group box transcription factor 1, HMG box transcription factor 1, HMG box-containing protein 1, HMG-box containing protein 1, HMG-box transcription factor 1, USMCC2 | |
| Produced in rabbits immunized with purified, recombinant Human HBP1 (Accession#: O60381-1; Pro208-Phe345) | |
| 0.1 mL | |
| Cardiovascular Biology, Stem Cells | |
| 26959 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction