missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBO1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38191-20ul
This item is not returnable.
View return policy
Description
HBO1 Polyclonal antibody specifically detects HBO1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| HBO1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| HBO1EC 2.3.1.48, HBOA, Histone acetyltransferase binding to ORC1, histone acetyltransferase MYST2, KAT7, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 2, MYST histone acetyltransferase 2, MYST-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 55-193 of human HBO1 (NP_008998.1).,, Sequence:, DSSPVRNLQSFGTEEPAYSTRRVTRSQQQPTPVTPKKYPLRQTRSSGSETEQVVDFSDRETKNTADHDESPPRTPTGNAPSSESDIDISSPNVSHDESIAKDMSLKDSGSDLSHRPKRRRFHESYNFNMKCPTPGCNSL | |
| 20 μL | |
| Cell Cycle and Replication | |
| 11143 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction