missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HBO1 Antibody (4E12H12), Novus Biologicals™

Mouse Monoclonal Antibody

£217.00 - £389.00

Specifications

Antigen HBO1
Clone 4E12H12
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30034922
View Documents
Novus Biologicals™
NBP3-27149-0.025ml
0.025 mL
£217.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30037668
View Documents
Novus Biologicals™
NBP3-27149-0.1ml
0.1 mL
£389.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HBO1 Monoclonal antibody specifically detects HBO1 in Human,Monkey,Mouse samples. It is validated for ELISA,Western Blot
Specifications

Specifications

HBO1
1 mg/mL
ELISA, Western Blot
Unconjugated
Mouse
Cell Cycle and Replication
PBS
11143
IgG1
Protein G purified
4E12H12
Western Blot 1:500 - 1:2000, ELISA 1:10000
Monoclonal
Purified
RUO
Human, Monkey, Mouse
HBO1EC 2.3.1.48, HBOA, Histone acetyltransferase binding to ORC1, histone acetyltransferase MYST2, KAT7, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 2, MYST histone acetyltransferase 2, MYST-2
Purified recombinant fragment of human HBO1 (AA: 1-200) expressed in E. Coli.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.