missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBG1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19993-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to HBG1 and HBG2. The antibody has no cross reactivity with HBB and HBD.
Hemorrhage of cells from an Rh+ fetus into the circulation of an Rh- mother may result in the formation of Rh-reactive antibodies in the mother. Rh hemolytic anemia in a subsequent Rh+ fetus may result from placental transfer of antibodies formed in the mother to the fetus. Although the volume of fetal erythrocytes found in the maternal circulation during pregcy and immediately post-partum is reported to be small and of uncertain clinical significance in many cases, substantial hemorrhage may result from a number of causes including fetal or maternal trauma and placental defects. Erythrocytes containing fetal hemoglobin may be found in individuals of any age, but with lower amounts of fetal hemoglobin compared to fetal red cells. These cells have been termed F cells. High levels of F cells may also exist in adults with a heterogeneous group of genetic disorders of uncertain etiology, referred to as Hereditary Persistence of Fetal Hemoglobin. Other clinical conditions causing significant levels of anemia may also result in elevated levels of F cells. Several clinical conditions have been described with increased levels of F cells. These conditions include hereditary anemic diseases such as sickle cell anemia and thalassemia major.Specifications
| HBG1 | |
| Polyclonal | |
| Unconjugated | |
| HBG2 | |
| Gamma 1 globin, Hb F Agamma, HBG1, HBGA, HBGR, Hemoglobin gamma 1 chain, Hemoglobin gamma A chain, Hemoglobin subunit gamma 1, hemoglobin, gamma A, HSGGL1, PRO2979 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3047, 3048 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P69891, P69892 | |
| HBG1, HBG2 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction