missing translation for 'onlineSavingsMsg'
Learn More

HBG1 Rabbit anti-Human, Clone: 1R9T7, Novus Biologicals™

Product Code. 18302114 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18302114 20 μg 20µL
18355714 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18302114 Supplier Novus Biologicals Supplier No. NBP31529820UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HBG1 Monoclonal antibody specifically detects HBG1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBG1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 1R9T7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias gamma-1-globin, Hb F Agamma, HBGA, HBGR, HBG-T2, Hemoglobin gamma A, Hemoglobin gamma-1 chain, Hemoglobin gamma-A chain, Hemoglobin subunit gamma 1, HSGGL1, PRO2979
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 68-147 of human HBG1 (P69891).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3047
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.