missing translation for 'onlineSavingsMsg'
Learn More

HBG1/2 Antibody (1G8), Novus Biologicals™

Product Code. 18325937 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325937 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325937 Supplier Novus Biologicals Supplier No. H00003047M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HBG1/2 Monoclonal antibody specifically detects HBG1/2 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBG1/2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1G8
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH10914
Gene Alias abnormal hemoglobin, A-gamma globin, gamma A hemoglobin, gamma globin, Gamma-1-globin, gamma-2-globin, gamma-globin chain, G-gamma globin Paulinia, Hb F Agamma, hb F Ggamma, HBGA, HBGR, HBG-T1, HBG-T2, Hemoglobin gamma-1 chain, hemoglobin gamma-2 chain, Hemoglobin gamma-A chain, hemoglobin gamma-G chain, hemoglobin subunit gamma-1, hemoglobin subunit gamma-2, hemoglobin, gamma A, hemoglobin, gamma G, hemoglobin, gamma, regulator of, HSGGL1, methemoglobin, PRO2979, TNCY
Host Species Mouse
Immunogen HBG1 (AAH10913, 1 a.a. ~ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGHFTEEDKATITSLWGKVNVEDAGGETLGRLLVVYPWTQRFFDSFGNLSSASAVMGNPKVKAHGKKVLTSLGDAIKHLDDLKGTFAQLSELHCDKLHVDPENFKLLGNVLVTVLAIHFGKEFTPEVQASWQKMVTGVASALSSRYH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 3047
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.