missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBEGF, Mouse anti-Human, Clone: 2D10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00001839-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: LVTGESLERLRRGLAAGTSNPDPPTVSTDQLLPLGGGRDRKVRDLQEADLDLLRVTLSSKPQALATPNKEEHGKRKKKGKGLGKKRDPCLRKYKDFCIHGECKYVKELRAPSCICHPGYHGERCHGLSLPVENRLYTYDHTTILAVVAVVLSSVCLLVIVGLLMFRYHRRGGYDVENEEKVKLGMTNSHSpecifications
| HBEGF | |
| Monoclonal | |
| Unconjugated | |
| heparin-binding EGF-like growth factor | |
| DTR/DTS/DTSF/HEGFL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2D10 | |
| In 1x PBS, pH 7.4 | |
| BC033097 | |
| HBEGF | |
| HBEGF (AAH33097.1, 20 a.a. ∼ 208 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 1839 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction