missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HBE1 Recombinant Rabbit Monoclonal Antibody (HL2457)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550538
This item is not returnable.
View return policy
Description
HBE1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The epsilon globin gene (HBE) is normally expressed in the embryonic yolk sac: two epsilon chains together with two zeta chains (an alpha-like globin) constitute the embryonic hemoglobin Hb Gower I; two epsilon chains together with two alpha chains form the embryonic Hb Gower II. Both of these embryonic hemoglobins are normally supplanted by fetal, and later, adult hemoglobin. The five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5'-epsilon - G-gamma - A-gamma - delta - beta-3'.
Specifications
| HBE1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P02100 | |
| HBE1 | |
| Synthetic peptide of human Hemoglobin epsilon. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2457 | |
| Unconjugated | |
| HBE1 | |
| A-gamma globin; Beta-4 globin; epsilon 1 globin; epsilon globin; epsilon I globin; Epsilon-1-globin; Epsilon-globin; GLNB1; globin B1; HBE; HBE1; HBG1; HBG2; Hemoglobin epsilon chain; Hemoglobin epsilon-1 chain; Hemoglobin epsilon-I chain; hemoglobin subunit epsilon; hemoglobin subunit epsilon 1; Hemoglobin subunit epsilon-1; hemoglobin subunit epsilon-1-like; hemoglobin, epsilon 1; hemoglobin, gamma A; hemoglobin, gamma G; LOC102174495 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 110337495, 293267, 3046 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction