Learn More
Invitrogen™ HBA1/HBA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143842
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, mouse heart tissue. IHC: human gastric signet ring cell carcinoma tissue, human rectal moderately differentiated adenocarcinoma tissue, human spleen tissue, human renal cancer tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.
Specifications
| HBA1/HBA2 | |
| Polyclonal | |
| Unconjugated | |
| Hba1 | |
| alpha globin; alpha one globin; alpha-2 globin; alpha-2 globin chain; alpha-globin; delta globin; HBA1; HBA2; HBA-T2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin subunit alpha 2; hemoglobin, alpha 1; hemoglobin, alpha 2; mutant hemoglobin alpha 2 globin chain | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3039, 3040 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P69905 | |
| Hba1, Hba2 | |
| A synthetic peptide corresponding to a sequence of human Hemoglobin/HBA1/HBA2 (AVHASLDKFLASVSTVLTSKYR). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.