missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HB9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20975-1-AP-20UL
This item is not returnable.
View return policy
Description
Homeobox protein HB9 is expressed in lymphoid and pancreatic tissues. It is reported to be Putative transcription factor involved in pancreas development and function. Defects in this gene causes Currarino syndrome. The triad of a presacral tumor, sacral agenesis and anorectal malformation constitutes the Currarino syndrome which is caused by dorsal-ventral patterning defects during embryonic development. It is also reported that HB9 is strongly expressed in epidermal basal cells of the outer scale face in tarsometatarsal scale skin.Specifications
| HB9 | |
| Polyclonal | |
| Unconjugated | |
| MNX1 | |
| HB9, HLXB9, Homeobox protein HB9, HOXHB9, MNX1, SCRA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15285, 3110 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50219, Q9QZW9 | |
| Mnx1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion