missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HB-EGF Antibody (KHK2866) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28182-100ug
This item is not returnable.
View return policy
Description
HB-EGF Monoclonal antibody specifically detects HB-EGF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| HB-EGF | |
| Monoclonal | |
| Unconjugated | |
| Q99075 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG |
| Flow Cytometry, ELISA, Functional Assay | |
| KHK2866 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor | |
| HBEGF | |
| 100 μg | |
| Biologically Active Proteins | |
| 1839 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction