missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Pig™ HazMat Polypropylene Chemical Absorbent Pulp
Click to view available options
Packaging Quantity:
1 Ea.
Unit Size:
Each
Specifications
Specifications
| Color | Pink |
| Material | Polypropylene |
| Type | Hazmat Chemical Absorbent Pulp |
| For Use With (Application) | Ideal for difficult to reach spaces or final cleanup |
| Packaging Quantity | 1 Ea. |
| Weight (English) | 5 lb. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction