missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Hazardous Substances Rack

Product Code. p-200003650 Shop All asecos Products
Click to view available options
Capacity (Metric):
30 L
40 L
Weight (Metric):
56 kg
59 kg
64 kg
71 kg
90 kg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17816376

Brand: asecos™ 27853

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The Hazardous Substances Rack provides storage of aggressive chemicals.

  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • Varioius storage level options
  • Polyethylene sumps with high resistance against aggressive chemicals (such as acids and alkalis)

Specifications

Capacity (English) 7.9 gal.
Capacity (Metric) 30 L
Material Steel
Type Drum Rack
Length (English) 41.73 in.
Length (Metric) 1060.00 mm
Width (English) 41.73 in.
Width (Metric) 1060.00 mm
Height (English) 78.74 in.
Height (Metric) 2000.00 mm
Depth (English) 25.08 in.
Depth (Metric) 637.00 mm
Weight (English) 123.46 lb.
Weight (Metric) 56 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.