Learn More
Key Industrial Equipment™ Hazardous storage cabinet, for acid/alkali, white, 610mm x 459mm x 459mm
Brand:  Key Industrial Equipment™ A350586
Encompass
Product Code. 12891517
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Hard wearing cabinets suitable for storing mild acids and corrosive chemicals.
  • One shelf
  • Dimensions (H x W x D) : 610mm x 459mm x 459mm