missing translation for 'onlineSavingsMsg'
Learn More

3M™ Hazardous Spill Response Kit

Product Code. 12372459
Change view
Click to view available options
Capacity (Metric):
5 L
26 L
Dimensions (L x W):
400 x 70 mm
500 x 630 mm
Includes:
10 sheets, 1 pillow, 1 disposable bag with tie
10 sheets, 5 pillows, 5 mini-booms and 2 disposable bags with ties.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Dimensions (L x W) Includes unitSize
12372459 5 L 400 x 70 mm 10 sheets, 1 pillow, 1 disposable bag with tie Each
12327979 26 L 500 x 630 mm 10 sheets, 5 pillows, 5 mini-booms and 2 disposable bags with ties. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12372459 Supplier 3M™ Supplier No. HSRK5

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Contains chemical sorbents made from inert, synthetic fibers to be deployed wherever hazardous liquids are in use, for the protection of both employees and equipment. 3M™ Hazardous Spill Response Kits can be used to contain and clean up the hazardous liquid without delay.

Kits contain chemical sorbents made from inert, synthetic fibers to be deployed wherever hazardous liquids are in use, for the protection of both employees and equipment. In the event of a spill, the kit can be used to contain and clean up the hazardous liquid without delay.

  • Constructed from polypropylene fibers
  • Available in a wide range of formats
  • Lightweight and dust-free
  • High absorption capacity minimizes waste for disposal.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5 L
Material Polypropylene Fiber
Type Spill Kit
Includes 10 sheets, 1 pillow, 1 disposable bag with tie
Packaging Quantity 1 Ea.
Dimensions (L x W) 400 x 70 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.