missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Hazardous Chemical Labels for B30 Series

Product Code. p-7225698
Click to view available options
Display:
L3B Layout
L4A Layout
Unit Size:
Pack of 200
This item is not returnable. View return policy

Product Code. 15931011

Brand: Brady™ 121649

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pre-printed, GHS/CLP chemical labels for BBP3X, S3XXX, and i3300 printers.

  • Permanent acrylic adhesive
  • For smooth and textured surfaces

Specifications

Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Color Red on White
Display L4A Layout
Finish Gloss
Material Vinyl (B-7569)
Print Technology Thermal Transfer
Sterility Non-sterile
For Use With (Application) Facility Identification, Hazard Communication Identification, Electrical Labeling
For Use With (Equipment) BBP™30, BBP™31, BBP™33, BBP™35 Multicolor, BBP™37 Multicolor, BradyPrinter i3300, BradyPrinter S3000, S3100, and i5300 Industrial Label Printers
Labels per Row 1
Quantity 200 Labels
Shape Rectangle
Temperature Range (Metric) -20°C to +75°C
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 162 mm
Width (Metric) 155 mm
Height (Metric) 105 mm
Alternate Part Number B30-262-7569-CLP4A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.