Learn More
Hazard Warning Label 'Oxidiser' Self-Adhesive GHS and CLP Compliant 20x20mm
Product Code. 12829768
Encompass_Preferred
Product Code. 12829768 Supplier Brand Not Defined Supplier No. F028 01 025
Only null left
Add to basket
Add to basket
All hazard warning diamonds are laminated and produced on a polypropylene material. The base material used conforms with B.S 5609:1986 which is perfect for transport by Road, Rail, Air and Sea. Hazard warning diamonds fully comply with the GHS, CLP, RID IMDG/IMO, ADN and ICAO/IATA requirements.
They have the following main properties:
• Excellent Weather Resistance.
• Excellent Chemical Resistance.
• Excellent Abrasion Resistance.
• Marine Grade Adhesive