missing translation for 'onlineSavingsMsg'
Learn More

New Pig Haz-Mat Socks

Stop the spread of acids and bases.

Brand:  New Pig PIG301

Code : Z4

Additional Details : Weight : 6.80000kg

 View more versions of this product

Product Code. 12449277

  • £162.69 / Pack of 6
Estimated Shipment: 17-05-2024
to see stock.
Add to basket

Description

Description

  • Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Excellent for quickly diking and stopping the spread of chemical spills
  • Absorb and retain most acids, bases and unknown liquids
  • Can be incinerated after use to reduce waste

Specifications

Specifications

Pink
7.6 L
Hazmat Absorbent Sock
10 ft. L x 3 in. W (3.04 m x 7.62 cm)
10 ft.
3 in.
3 in.
14 lb.
2 gal.
Polypropylene
Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives
10 ft. x 3 in. (3.04 m x 7.62 cm)
3.04 m
7.62 cm
7.62 cm
6.4 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers