Learn More
Cole-Parmer™ Hayward PVC Needle Valve
Product Code. 11825342
Change view
Click to view available options
Thread Size:
3/8 in. NPT(F)
Unit Size:
Each
Product Code. Thread Size unitSize
11825342 3/8 in. NPT(F) Each
1 options
Product Code. 11825342 Supplier Cole-Parmer™ Supplier No. 0324562
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Precision needle valves feature an integrated stem with PTFE seat; fine pitch stem threads ensure precise adjustment.
  • Accurate flow control lets you adjust flow down to drops per minute
  • Seals are heavy-duty FPM O-ring
  • Natural PVDF valves are the ideal option for high-purity, high-temperature applications
  • Valve body includes flanges for panel mounting
  • Connections are standard pipe thread
Specifications
Material PVC, PTFE, Viton
Product Line Hayward 155.522.554 PVC Needle Valve
Inlet 3/8 in. NPT(F)
Outlet Port 3/8 in. NPT(F)
Port Size 0.375 NPT(F)
Thread Size 3/8 in. NPT(F)
Material (Seal) FPM O-ring
Maximum Pressure 150 psi
Max. Operating Temperature (English) 140°F
Max. Operating Temperature (Metric) 60°C
Product Type PVC Needle Valve