missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HaXS8
GREENER_CHOICE

Product Code. 18732138
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18732138 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18732138 Supplier Tocris Bioscience™ Supplier No. 4991/2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical dimerizer

HaXS8 is a promotes covalent and irreversible HaloTag™ and SNAP-tag™ substrate dimerization. Cell permeable.

HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[[4-[[(2-Amino-9-H-purin-6-yl)oxy]methyl]phenyl]methyl]-2-[4-[(18-chloro-3,6,9,12-tetraoxaoctadec-1yl)oxy]-2,3,5,6-tetrafluorophenoxy]acetamide
CAS 2080306-25-6
Quantity 2 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C35H43ClF4N6O8
Formula Weight 787.2
Product Type Chelator

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.