missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAX1 Recombinant Rabbit Monoclonal Antibody (HL2170)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550416
This item is not returnable.
View return policy
Description
HAX1 Recombinant Monoclonal Antibody for IHC (P), Western Blot
The protein encoded by this gene is known to associate with hematopoietic cell-specific Lyn substrate 1, a substrate of Src family tyrosine kinases. It also interacts with the product of the polycystic kidney disease 2 gene, mutations in which are associated with autosomal-dominant polycystic kidney disease, and with the F-actin-binding protein, cortactin. It was earlier thought that this gene product is mainly localized in the mitochondria, however, recent studies indicate it to be localized in the cell body. Mutations in this gene result in autosomal recessive severe congenital neutropenia, also known as Kostmann disease. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| HAX1 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with no preservative | |
| O00165, O35387 | |
| Hax1 | |
| Recombinant fragmemt of human HAX1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2170 | |
| Unconjugated | |
| Hax1 | |
| FLJ17042; FLJ18492; FLJ93803; Hax1; HAX-1; HCLS1 (and PKD2) associated protein; HCLS1 associated protein X-1; HCLS1 associated X-1; HCLS1-associated protein X-1; HCLSBP1; HS1 binding protein; HS1-associated protein X-1; HS1-associating protein X-1; HS1-binding protein 1; HS1BP1; HSP1BP-1; mHAX-1s; OTTHUMP00000034190; RP11-137P24.1; SCN3; SIG-111; Silg111 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10456, 23897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction