missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HAX1 Monoclonal Antibody (8F9G7)
GREENER_CHOICE

Product Code. 12867792
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12867792 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12867792 Supplier Invitrogen™ Supplier No. MA515769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HAX1 Monoclonal Antibody for Western Blot

The protein encoded by this gene is known to associate with hematopoietic cell-specific Lyn substrate 1, a substrate of Src family tyrosine kinases. It also interacts with the product of the polycystic kidney disease 2 gene, mutations in which are associated with autosomal-dominant polycystic kidney disease, and with the F-actin-binding protein, cortactin. It was earlier thought that this gene product is mainly localized in the mitochondria, however, recent studies indicate it to be localized in the cell body. Mutations in this gene result in autosomal recessive severe congenital neutropenia, also known as Kostmann disease. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAX1
Applications Western Blot
Classification Monoclonal
Clone 8F9G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Hax1
Gene Accession No. O00165, O35387, Q7TSE9
Gene Alias FLJ17042; FLJ18492; FLJ93803; Hax1; HAX-1; HCLS1 (and PKD2) associated protein; HCLS1 associated protein X-1; HCLS1 associated X-1; HCLS1-associated protein X-1; HCLSBP1; HS1 binding protein; HS1-associated protein X-1; HS1-associating protein X-1; HS1-binding protein 1; HS1BP1; HSP1BP-1; mHAX-1s; OTTHUMP00000034190; RP11-137P24.1; SCN3; SIG-111; Silg111
Gene Symbols Hax1
Host Species Mouse
Immunogen A 15 amino acid peptide near the amino terminus of human Hax1a.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10456, 23897, 291202
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.