missing translation for 'onlineSavingsMsg'
Learn More

HAX-1 Antibody [DyLight 594], Novus Biologicals™

Product Code. 30737896 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737896 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737896 Supplier Novus Biologicals Supplier No. AF5458M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

HAX-1 Polyclonal antibody specifically detects HAX-1 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAX-1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias FLJ17042, FLJ93803, HAX-1, HCLS1 (and PKD2) associated protein, HCLS1 associated protein X-1, HCLS1-associated protein X-1, HCLSBP1, HS1 binding protein, HS1-associating protein X-1, HS1-binding protein 1, HS1BP1FLJ18492, HSP1BP-1, SCN3
Host Species Goat
Immunogen E. coli-derived recombinant human HAX-1, Ser2-Leu261, Accession # O00165
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10456
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.