missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HAX-1 Polyclonal antibody specifically detects HAX-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | HAX-1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ17042, FLJ93803, HAX-1, HCLS1 (and PKD2) associated protein, HCLS1 associated protein X-1, HCLS1-associated protein X-1, HCLSBP1, HS1 binding protein, HS1-associating protein X-1, HS1-binding protein 1, HS1BP1FLJ18492, HSP1BP-1, SCN3 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human Hax1a. The immunogen is located within the first 50 amino acids of Hax1a. Amino Acid Squence: GPRPSHRDPFFGGMTRD |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?