missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HAVCR2 Recombinant Protein

Product Code. 16186123
Click to view available options
Quantity:
10 μg
25 μg
missing translation for 'unitSize'
10µg
25µg
This item is not returnable. View return policy

Product Code. 16186123

missing translation for 'mfr': Abnova™ H00084868P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human HAVCR2 full-length ORF recombinant protein with GST-tag at N-terminal

CD4-positive T helper lymphocytes can be divided into types 1 (Th1) and 2 (Th2) on the basis of their cytokine secretion patterns. Th1 cells and their associated cytokines are involved in cell-mediated immunity to intracellular pathogens and delayed-type hypersensitivity reactions, whereas Th2 cells are involved in the control of extracellular helminthic infections and the promotion of atopic and allergic diseases. The 2 types of cells also cross-regulate the functions of the other. TIM3 is a Th1-specific cell surface protein that regulates macrophage activation and enhances the severity of experimental autoimmune encephalomyelitis in mice.

  • Theoretical MW (kDa): 41.36
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MFSHLPFDCVLLLLLLLLTRSSEVEYRAEVGQNAYLPCFYTPAAPGNLVPVCWGKGACPVFECGNVVLRTDERDVNYWTSRYWLNGDFRKGDVSLTIENVTLADSGIYCCRIQIPGIMNDEKFNLKLVIKPGEWTFACHLYE

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH20843
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 84868
Molecular Weight (g/mol) 41.36
Name HAVCR2 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Source Wheat Germ (in vitro)
Immunogen MFSHLPFDCVLLLLLLLLTRSSEVEYRAEVGQNAYLPCFYTPAAPGNLVPVCWGKGACPVFECGNVVLRTDERDVNYWTSRYWLNGDFRKGDVSLTIENVTLADSGIYCCRIQIPGIMNDEKFNLKLVIKPGEWTFACHLYE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FLJ14428/KIM-3/TIM3/TIMD3/Tim-3
Common Name HAVCR2
Gene Symbol HAVCR2
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.