missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAUS3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5117979
This item is not returnable.
View return policy
Description
HAUS3 Polyclonal antibody specifically detects HAUS3 in Human samples. It is validated for Immunoprecipitation, Western Blot
Specifications
| HAUS3 | |
| Polyclonal | |
| PBS with 0.03% Proclin 300; pH 7 | |
| Q68CZ6 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 79441 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| HAUS3 | |
| C4orf15; D4S43h; D5H4S43; D5H4S43E; dgt3; HAUS augmin like complex subunit 3; HAUS augmin-like complex subunit 3; HAUS augmin-like complex, subunit 3; HAUS3; IT1 | |
| E. coli-derived Human HAUS3 fragment | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction