missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAUS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565275
This item is not returnable.
View return policy
Description
Immunogen sequence: IHEVVESSNED NFQLLDIQTP SICDNQEILE ERRLEMARLQ LAYICAQHQL IHLKASNSSM KSSIKWAEES LHSLTSKAVD KENLDAKISS LTSEIM Highest antigen sequence identity to the following orthologs - mouse 78%, rat 84%.
Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
Specifications
| HAUS3 | |
| Polyclonal | |
| Unconjugated | |
| HAUS3 | |
| C4orf15; D4S43h; D5H4S43; D5H4S43E; dgt3; HAUS augmin like complex subunit 3; HAUS augmin-like complex subunit 3; HAUS augmin-like complex, subunit 3; HAUS3; IT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79441 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68CZ6 | |
| HAUS3 | |
| Recombinant Human HAUS3. Recombinant protein control fragment (Product #RP-105991). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction