missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: MSCGNEFVET LKKIGYPKAD NLNGEDFDWL FEGVEDESFL KWFCGNVNEQ NVLSERELEA FSILQKSGKP ILEGAALDEA LKTCKTSDLK TPRLDDKELE KLEDEVQTLL KLKNLK Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 82%.
Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
Specifications
Specifications
| Antigen | HAUS3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HAUS3 |
| Gene Accession No. | Q68CZ6 |
| Gene Alias | C4orf15; D4S43h; D5H4S43; D5H4S43E; dgt3; HAUS augmin like complex subunit 3; HAUS augmin-like complex subunit 3; HAUS augmin-like complex, subunit 3; HAUS3; IT1 |
| Gene Symbols | HAUS3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
