missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HAUS2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA559013

Product Code. 15756881

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IPHLAANLKK MNQALAKMDI LVTETEELAE NILKWRKQQN EVSSCIPKIL AEESYLYKHD IIMPPLPFTS KVHVQTINAK Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 65%.

HAUS2 is 1 of 8 subunits of the 390-kD human augmin complex, or HAUS complex. The augmin complex was first identified in Drosophila, and its name comes from the Latin verb augmentare, meaning to increase. The augmin complex is a microtubule-binding complex involved in microtubule generation within the mitotic spindle and is vital to mitotic spindle assembly (Goshima et al., 2008 [PubMed 18443220]; Uehara et al., 2009 [PubMed 19369198]).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HAUS2
Polyclonal
Unconjugated
HAUS2
C15orf25; centrosomal protein 27kDa; centrosomal protein of 27 kDa; Cep27; HAUS augmin like complex subunit 2; HAUS augmin-like complex subunit 2; HAUS augmin-like complex, subunit 2; HAUS2; HsT17025
Rabbit
Antigen affinity chromatography
RUO
55142
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.7 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NVX0
HAUS2
Recombinant protein corresponding to Human HAUS2. Recombinant protein control fragment (Product #RP-98069).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.