missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581190
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human KAT1/HAT1.
The protein encoded by this gene is a type B histone acetyltransferase (HAT) that is involved in the rapid acetylation of newly synthesized cytoplasmic histones, which are in turn imported into the nucleus for de novo deposition onto nascent DNA chains. Histone acetylation, particularly of histone H4, plays an important role in replication-dependent chromatin assembly. Specifically, this HAT can acetylate soluble but not nucleosomal histone H4 at lysines 5 and 12, and to a lesser degree, histone H2A at lysine 5. Alternatively spliced transcript variants have been identified for this gene.
Specifications
| HAT1 | |
| Polyclonal | |
| Unconjugated | |
| HAT1 | |
| 2410071B14Rik; AA536933; HAT1; Hat-1; histidine aminotransferase 1; histone acetyltransferase 1; Histone acetyltransferase type B catalytic subunit; histone aminotransferase 1; KAT1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 8520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| O14929 | |
| HAT1 | |
| A synthetic peptide corresponding to the N-terminus of the Human HAT1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction