Learn More
Hastelloy C∣r gauze, 40 mesh woven from 0.19mm (0.0075in) dia wire
Description
Hastelloy C is used in combustion chambers, heat-treating equipment, collector rings, combustion chambers and exhaust stacks for high-heat applications in for conventional aircraft and jets, pipe connections, pumps and valves for oil refinery, chemical and petrochemical applications. Also used in components exposed to flue gas or flue gas desulfurization plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Hastelloy C™ gauze |
| Name Note | 40 mesh woven from 0.19mm (0.0075 in.) dia. wire |
| Quantity | 150 x 150 mm |
| Mesh Size | 40 Mesh |
| Form | Wire Cloth |
| MDL Number | MFCD00801065 |
| Solubility Information | Insoluble in water. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.