missing translation for 'onlineSavingsMsg'
Learn More

Hastelloy C∣r gauze, 20 mesh woven from 0.23mm (0.009in) dia wire

Product Code. 11365247
Change view
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365247 300 x 300 mm Each
11385247 75 x 75 mm Each
11375247 150 x 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365247 Supplier Thermo Scientific Alfa Aesar Supplier No. 040938.HB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High strength, corrosion resistant alloy

Hastelloy C™ gauze is used as high strength, corrosion resistant alloy. It is used in a range of industries such as chemical processing, aerospace, pollution-control equipment, marine engineering.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hastelloy C™ gauze
Name Note Woven from 0.23mm (0.009 in.) dia. wire
Quantity 300 x 300 mm
Mesh Size 20 Mesh
Form Wire Cloth
MDL Number MFCD00801065
Synonym Nickel Molybdenum Chromium
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.