missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Hastelloy C™ gauze, 100 mesh woven from 0.102mm (0.004 in.) dia. wire

Brand:  Alfa Aesar™ 040940.HB

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11345257

  • £279.00 / Each


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Specifications

Hastelloy C™ gauze
300 x 300mm
Wire Cloth
100 Mesh
Woven from 0.102mm (0.004 in.) dia. wire
MFCD00801065
Videos
SDS