missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15609-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
HAS1, HAS2 and HAS3 are HA (hyaluro or hyaluronic acid) synthase proteins. The extracellular matrix in most vertebrates express HA, which is a high molecular weight linear polysaccharide composed of alternating glucuronic acid and N-acetylglucosamine residues linked by b-1,3 and b-1,4 glycosidic bonds. The three HAS genes show distinct patterns of expression during development and their protein products play significantly different roles in the formation of the HA matrix. Both HAS1 and HAS2 synthesize high molecular-weight HA, whereas HAS3 produces lower molecular weight HA. The expression of the three HAS isoforms is more prominent in growing cells than in resting cells and is differentially regulated by various stimuli, suggesting distinct functional roles of the three proteins. HAS3 produces both secreted and cell-associated forms of hyaluro and is the most active of the three isoforms of this enzyme in adults. HAS3 gene expression plays a crucial role in the regulation of hyaluro synthesis in the epidermis. Specifically, IFN-g markedly upregulates HAS3 mRNA, whereas TGFbeta downregulates HAS3 transcript levels. The human HAS3 gene maps to chromosome 16q22.1.Specifica
| HAS3 | |
| Polyclonal | |
| Unconjugated | |
| HAS3 | |
| HA synthase 3, HAS3, hyaluro synthase 3, Hyaluronate synthase 3, Hyaluronic acid synthase 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15118, 266805, 3038 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00219, O08650 | |
| Has3 | |
| HAS3 Fusion Protein Ag7999 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto