Learn More
Invitrogen™ HAS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589266
Description
Immunogen sequence: HRRMRRAGQA LKLPSPRRGS VALCIAAYQE DPDYLRKCLR SAQRISFPDL KVVMVVDGNR QEDAYMLDIF HEVLGGTEQA GFFVWRSNFH EAGEGETEAS LQEGMDRVRD VVRASTFSCI MQKWGGKREV MYTAFKALGD SVDYIQVCDS DTVLDPACTI EMLRVLEEDP QVGGVGGDVQ PPGKGMAVED DQVQAAQVRA TEAWSVHQRH VSREQ; Positive Samples: SKOV3, SW480, Mouse heart, Rat heart, Rat liver; Cellular Location: Membrane, Multi-pass membrane protein.
HAS1, HAS2 and HAS3 are HA (hyaluronan or hyaluronic acid) synthase proteins. The extracellular matrix in most vertebrates express HA, which is a high molecular weight linear polysaccharide composed of alternating glucuronic acid and N-acetylglucosamine residues linked by b-1,3 and b-1,4 glycosidic bonds. The three HAS genes show distinct patterns of expression during development and their protein products play significantly different roles in the formation of the HA matrix. Both HAS1 and HAS2 synthesize high molecular-weight HA, whereas HAS3 produces lower molecular weight HA. The expression of the three HAS isoforms is more prominent in growing cells than in resting cells and is differentially regulated by various stimuli, suggesting distinct functional roles of the three proteins. HAS3 produces both secreted and cell-associated forms of hyaluronan and is the most active of the three isoforms of this enzyme in adults. HAS3 gene expression plays a crucial role in the regulation of hyaluronan synthesis in the epidermis. Specifically, IFN-g markedly upregulates HAS3 mRNA, whereas TGFbeta downregulates HAS3 transcript levels. The human HAS3 gene maps to chromosome 16q22. 1.
Specifications
| HAS3 | |
| Polyclonal | |
| Unconjugated | |
| HAS3 | |
| HA synthase 3; Has3; Hyaluronan synthase 3; hyaluronate synthase 3; Hyaluronic acid synthase 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15118, 266805, 3038 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00219, O08650 | |
| HAS3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 67-281 of human HAS3 (NP_6195151). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.