missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAS2 Monoclonal Antibody (2105CT879.18.75)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537783
This item is not returnable.
View return policy
Description
HAS2 Monoclonal Antibody for Western Blot
Hyaluronan or hyaluronic acid (HA) is a high molecular weight unbranched polysaccharide synthesized by a wide variety of organisms from bacteria to mammals, and is a constituent of the extracellular matrix. It consists of alternating glucuronic acid and N-acetylglucosamine residues that are linked by beta-1-3 and beta-1-4 glycosidic bonds. HA is synthesized by membrane-bound synthase at the inner surface of the plasma membrane, and the chains are extruded through pore-like structures into the extracellular space. It serves a variety of functions, including space filling, lubrication of joints, and provision of a matrix through which cells can migrate. HA is actively produced during wound healing and tissue repair to provide a framework for ingrowth of blood vessels and fibroblasts. Changes in the serum concentration of HA are associated with inflammatory and degenerative arthropathies such as rheumatoid arthritis. In addition, the interaction of HA with the leukocyte receptor CD44 is important in tissue-specific homing by leukocytes, and overexpression of HA receptors has been correlated with tumor metastasis. HAS2 is a member of the newly identified vertebrate gene family encoding putative hyaluronan synthases, and its amino acid sequence shows significant homology to glycosaminoglycan synthetase (DG42) from Xenopus laevis, and human and murine hyaluronan synthase 1.
Specifications
| HAS2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q92819 | |
| HAS2 | |
| KLH conjugated synthetic peptide between 133-166 amino acids from the Central region of human HAS2. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 2105CT879.18.75 | |
| Unconjugated | |
| HAS2 | |
| HA synthase 2; Has2; Hyaluronan synthase 2; hyaluronan synthase homolog; Hyaluronan synthase type 2; hyaluronate synthase 2; Hyaluronic acid synthase 2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 3037 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction