missing translation for 'onlineSavingsMsg'
Learn More

HAS1 Monoclonal Antibody (5C9D6)
GREENER_CHOICE

Product Code. 17400853 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17400853 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17400853 Supplier Thermo Scientific Supplier No. MA538534

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HAS1 Monoclonal Antibody (5C9D6)

HAS1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen HAS1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5C9D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Has1
Gene Accession No. Q61647, Q92839
Gene Alias HA synthase 1; HAS; Has1; huHAS1; Hyaluronan synthase 1; hyaluronan synthase1; hyaluronate synthase 1; Hyaluronic acid synthase 1
Gene Symbols Has1
Host Species Mouse
Immunogen Purified recombinant fragment of human HAS1 (aa: (74-399)) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15116, 3036
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.